Learn More
Gilson™ PIPETMAN Neo™ Starter Kits Available on GSA/VA Contract for Federal Government customers only.
Contains three PIPETMAN Neo™ pipets plus boxes of certified-quality PIPETMAN Tips and other useful accessories. Gilson™ PIPETMAN Neo™ Starter Kit is dedicated to laboratories which are about to renew their old set of pipets especially those performing protocols requiring intensive pipetting.
Manufacturer:  Gilson™ F167600
 View more versions of this product
Catalog No. F167600G
Add to cart
Description
Description
Contains three pipettes conveniently packaged with useful accessories:
  • Three PIPETMAN Neo™ Pipets: P20N, P200N, and P1000N
  • Two racks of certified-quality PIPETMAN DIAMOND Tips: D200 and D1000
  • Three SINGLE Pipette holder
  • One Gilson Guide to Pipetting
  • One Two-minute inspection poster
  • Also includes three Single™ pipette holders and a Gilson Guide to Pipetting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
P10N, P100N and P1000N pipette, plus boxes of-quality, PIPETMAN tip DFL10ST, DF100ST and DF1000ST, three Single™ pipette holders, a two minute inspection poster and Gilson guide to pipetteting
Pipetteman™ Neo™ starter kit