Learn More
Gilson™ PIPETMAN™ Repair and Calibration Level 2 Pipet Service
Service level meets the requirements of most labs
Specifications
Product Type Level 2 pipette calibration and repair service
Warranty and Service Offering 1 Year
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) No. of Channels Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) No. of Channels Price Quantity & Availability  
LVL2SISOG
View Documents
Gilson™
LVL2SISO
ISO 17025 Single channel pipet 1 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Service level 2 for single channel and multichannel pipets includes:
  • Full internal and external cleaning and decontamination
  • Visual and functional inspection
  • Repair, gravimetric adjustment and calibration
  • Leak test
  • Calibration label and a detailed calibration certificate for each pipet
  • Free Parts: Free seal and o-ring for most brands of single channel pipettes as well as a free tip holder if evaluation shows it needs to be replaced. All other replacement parts are extra
Data is collected with 4 weighings at the low and high volumes
Order Info
  • Send your pipettes to the Gilson Service Center of Excellence or choose Gilson InLab™ Service with the added convenience of technicians performing calibration and service right in your lab.
  • Customers must fill out and send service order form with their pipettes
Warranty and Services
1 year
Specifications
Specifications
Level 2 pipette calibration and repair service
1 Year