Learn More
Gilson™ PIPETMAN™ TOWERPACK™ Refill System Available on GSA/VA Contract for Federal Government customers only.
Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. Gilson™ PIPETMAN™ TOWERPACK™ Refill System is available in autoclavable and sterilized versions.
Manufacturer:  Gilson™ F167103
 View more versions of this product
Catalog No. F167103G
Add to cart
Description
Description
  • Color-coded racks to differentiate tip models and indicate the compatible Gilson™ pipet models
  • Available in autoclavable and sterilized versions
  • Fits Universal Reload box (Cat. No. F167100G, available separately)
  • Sterilized Tower Pack is individually wrapped in medical device-quality plastic to help protect tips from contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
D200 2 to 200μL 51mm L Pipetman™ Diamond™ pipet tips
Tower Pack™ Pipet Refill System
2 - 200μL