Learn More
Gilson™ Safe Aspiration Station Available on GSA/VA Contract for Federal Government customers only.
Vacuum-Controlled Aspiration System with small footprint
Manufacturer:  Gilson™ F110741
 View more versions of this product
Catalog No. F110741G
Add to cart
Description
Description
Closed, stand-alone unit designed to prevent contamination hazards.
  • Compact design with adjustable speed allowing quick aspiration
  • Always ready to use by simply pressing the handle button
  • Handle accommodates different types of adaptors to aspirate from a variety of vessels
  • Convenient coupling for quick bottle emptying
  • In-line hydrophobic filter traps aerosols
  • Connect two handles to the station with the Y-shape connector
  • Motor starts only when the vacuum pressure drops
  • Level detection prevents overflow
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Safe Aspiration Station
1/Case