Learn More
Gilson™ Safe Aspiration Station
Vacuum-Controlled Aspiration System with small footprint
$17.00
Specifications
Color Clear
Quantity 1/Cs.
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
F1077418G
View Documents
Gilson™
F1077418
Tubing Connector
Each for $17.00
Add to cart
F110742G
View Documents
Gilson™
F110742
Safe Aspiration Kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Closed, stand-alone unit designed to prevent contamination hazards.
  • Compact design with adjustable speed allowing quick aspiration
  • Always ready to use by simply pressing the handle button
  • Handle accommodates different types of adaptors to aspirate from a variety of vessels
  • Convenient coupling for quick bottle emptying
  • In-line hydrophobic filter traps aerosols
  • Connect two handles to the station with the Y-shape connector
  • Motor starts only when the vacuum pressure drops
  • Level detection prevents overflow
Specifications
Specifications
Clear
1/Cs.