Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GilZ Rat anti-Mouse, DyLight 488, Clone: CFMKG15, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200143G
Description
GilZ Monoclonal specifically detects GilZ in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
GilZ | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TSC22D3 | |
The immunogen for this antibody is GILZ. | |
100 μL | |
Cancer | |
1831 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry | |
CFMKG15 | |
Western Blot, Flow Cytometry | |
Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title