Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159485
Description
GIMAP5 Polyclonal specifically detects GIMAP5 in Human samples. It is validated for Western Blot.Specifications
GIMAP5 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ11296, GTPase, IMAP family member 5, hIAN5, HIMAP3, IAN4, IAN4L1IROD, IAN5, IAN-5, IMAP3GTPase IMAP family member 5, immune associated nucleotide 4 like 1, immune associated nucleotide 4 like 1 (mouse), immunity associated protein 3, Immunity-associated nucleotide 4-like 1 protein, Immunity-associated nucleotide 5 protein, Immunity-associated protein 3, inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Rat: 91%. | |
Human, Mouse, Rat, Equine, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96F15 | |
GIMAP5 | |
Synthetic peptides corresponding to GIMAP5(GTPase, IMAP family member 5) The peptide sequence was selected from the middle region of GIMAP5 (NP_060854). Peptide sequence CERRYCAFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQL. | |
100 μL | |
Immunology | |
55340 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction