Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP6 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306038F
Description
GIMAP6 Polyclonal specifically detects GIMAP6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GIMAP6 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686A01175, FLJ22690, GTPase IMAP family member 6, GTPase, IMAP family member 6, hIAN2, hIAN6, human immune associated nucleotide 2, IAN2, IAN-2, IAN6, IAN-6, immune associated nucleotide 2, immune associated nucleotide 6, Immunity-associated nucleotide 2 protein, Immunity-associated nucleotide 6 protein | |
Produced in rabbits immunized with E. coli-derived Human GIMAP6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
474344 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title