Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginger Oil, FCC, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
45 kg
Packaging:
Amber Glass Bottle
Drum
8007-08-7
8007-08-7

GI115, 8007-08-7

Spectrum™ Ginger Oil, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Refractive Index 1.488-1.494
Quantity 500 mL
CAS Max % 1