Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GINS1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549424

Catalog No. PA549424


Add to cart

Description

Description

The yeast heterotetrameric GINS complex is made up of Sld5 (GINS4; MIM 610611), Psf1, Psf2 (GINS2; MIM 610609), and Psf3 (GINS3; MIM 610610). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
Specifications

Specifications

Gins1
Polyclonal
PBS with 0.09% sodium azide
Q9CZ15
DNA replication complex GINS protein PSF1, GINS complex subunit 1, Gins1, Psf1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
69270
Western Blot
Unconjugated
Gins1
Liquid
GINS1
KLH conjugated synthetic peptide between 152-186 amino acids from the C-terminal region of Mouse Gins1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit