Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIP Mouse anti-Human, Janelia Fluor 549, Clone: 5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP105162JF549

Catalog No. NBP15162549


Add to cart

Description

Description

GIP Monoclonal antibody specifically detects GIP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GIP
5
Immunocytochemistry/Immunofluorescence
Purified
GIP
Synthetic human gastric inhibitory polypeptide coupled to carrier
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Binds human GIP. Does not cross-react with GLP-1 or -2, PACAP, glucagon or VIP. Cross-reactivity with porcine GIP has not been determined.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Mouse
IgG1 Kappa
0.1mL
Primary
2695
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit