Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550611

Catalog No. PA550611


Add to cart

Description

Description

The antibody detects endogenous levels of total GIPR / GIP Receptor protein.

This gene encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
Specifications

Specifications

GIPR
Polyclonal
Unconjugated
GIPR
Liquid
GIPR
Synthetic peptide corresponding to residues near the C terminal of human gastric inhibitory polypeptide receptor
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1.1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P48546, Q0P543
GIP-R
Rabbit
IgG
100 μL
-20°C
Primary
2696, 381853
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit