Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Mouse anti-Human, Allophycocyanin/Cy7, Clone: 591827, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals MAB82102APCCY7

Catalog No. M82102APCY7


Add to cart

Description

Description

GIPR Monoclonal antibody specifically detects GIPR in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

GIPR
591827
Flow Cytometry
Conjugated, Purified
GIPR
NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466)
Protein A or G purified
GPCR, Neuroscience
Primary
2696.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit