Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR, Mouse anti-Human,Mouse,Rat,Canine, Clone: Gipg013, Novus Biologicals™

Mouse Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen GIPR
Clone Gipg013
Applications ELISA, Immunofluorescence, Blocking Assay
Host Species Mouse
Target Species Human, Mouse, Rat, Canine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015267
View Documents
Novus Biologicals
NBP2810670.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015268
View Documents
Novus Biologicals
NBP2810670.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

GIPR Monoclonal antibody specifically detects GIPR in Human, Mouse, Rat, Canine Samples. It is validated for ELISA, Immunofluorescence, Blocking Assay.
Specifications

Specifications

GIPR
ELISA, Immunofluorescence, Blocking Assay
Human, Mouse, Rat, Canine
Primary
Gipg013
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit