Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550969
Description
GIT2 Polyclonal Antibody detects endogenous levels of GIT2 protein.
GTPase-activating protein for the ADP ribosylation factor family.
Specifications
GIT2 | |
Polyclonal | |
Unconjugated | |
GIT2 | |
1500036H07Rik; 5830420E16Rik; 6430510B20Rik; 9630056M03Rik; ARF GAP GIT2; ARF GTPase activating protein 2; ARF GTPase-activating protein GIT2; B230104M05Rik; CAT2; CAT-2; cool associated tyrosine phosphorylated-2; cool-associated, tyrosine phosphorylated protein 2; cool-interacting tyrosine-phosphorylated protein 2; G protein-coupled receptor kinase interacting ArfGAP 2; G protein-coupled receptor kinase-interactor 2; GIT ArfGAP 2; Git2; GRK-interacting protein 2; KIAA0148; mKIAA0148; Paxillin kinase linker; PKL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26431, 304546, 9815 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q14161, Q9JLQ2 | |
GIT2 | |
Synthesized peptide derived from the Internal region of human GIT2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction