Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gitoxigenin 98%, Acros Organics

Click to view available options
Quantity:
100mg
545-26-6
C23H34O5
390.52
MFCD00021174
PVAMXWLZJKTXFW-VQMOFDJESA-N
gitoxigenin,16beta-hydroxydigitoxigenin,unii-g36k2h8sme,g36k2h8sme,3beta,14,16beta-trihydroxy-5beta-card-20 22-enolide,16beta-hydroxydisitoxigenin,16-beta-hydroxydigitoxigenin,5-beta-card-20 22-enolide, 3-beta,14,16-beta-trihydroxy,3-3s,5r,8r,9s,10s,13r,14s,16s,17r-3,14,16-trihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta a phenanthren-17-yl-2h-furan-5-one,16.beta.-hydroxydigitoxigenin
348482
CHEBI:38105
3-[(3S,5R,8R,9S,10S,13R,14S,16S,17R)-3,14,16-trihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
CC12CCC(CC1CCC3C2CCC4(C3(CC(C4C5=CC(=O)OC5)O)O)C)O
545-26-6
C23H34O5
390.52
MFCD00021174
PVAMXWLZJKTXFW-VQMOFDJESA-N
gitoxigenin,16beta-hydroxydigitoxigenin,unii-g36k2h8sme,g36k2h8sme,3beta,14,16beta-trihydroxy-5beta-card-20 22-enolide,16beta-hydroxydisitoxigenin,16-beta-hydroxydigitoxigenin,5-beta-card-20 22-enolide, 3-beta,14,16-beta-trihydroxy,3-3s,5r,8r,9s,10s,13r,14s,16s,17r-3,14,16-trihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta a phenanthren-17-yl-2h-furan-5-one,16.beta.-hydroxydigitoxigenin
348482
CHEBI:38105
3-[(3S,5R,8R,9S,10S,13R,14S,16S,17R)-3,14,16-trihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
CC12CCC(CC1CCC3C2CCC4(C3(CC(C4C5=CC(=O)OC5)O)O)C)O
Infrared Spectrum Authentic
Quantity 100mg
Color Yellow
Specific Rotation Condition +30.60° (22°C c=0.8,CH3OH)
Assay Percent Range 98%
Merck Index 15, 4468
Formula Weight 390.51
Physical Form Flakes
Percent Purity ≥97.5%
Chemical Name or Material Gitoxigenin, 98%

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger