Learn More
GITR (TNFRSF18) Monoclonal Antibody (T9P4G4-D7), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA545911
Encompass
Catalog No. PIMA545911
Add to Cart
This item is not returnable. View return policy
Description
Description
GITR (TNFRSF18) Monoclonal antibody specifically detects GITR (TNFRSF18) in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GITR (TNFRSF18)
Monoclonal
1 mg/mL
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4
O35714, Q9Y5U5
Tnfrsf18
Synthetic peptide from the C-terminal of Human Tumor Necrosis Factor Receptor Superfamily, Member 18
100 μg
Primary
Human, Mouse
Liquid
ELISA, Immunohistochemistry, Western Blot
T9P4G4-D7
PerCP
Tnfrsf18
Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364
Mouse
Protein G
RUO
21936, 8784
4° C
IgG2b
Recommendations
Recommendations