Learn More
GITR/TNFRSF18 Rat anti-Mouse, Alexa Fluor™ 350, Clone: DTA-1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP226661AF350
Catalog No. NP226661A30
Add to cart
Description
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
GITR/TNFRSF18
DTA-1
Flow Cytometry
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Rat
IgG2b Lambda
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TNFRSF18
Mouse CD25+ CD4+ T cell line NP_683700
0.1 mL
Primary
8784