Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITRLigand/TNFSF18 Mouse anti-Human, Janelia Fluor 646, Clone: MM0312-4Q25, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211965JF646
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry.Specifications
GITRLigand/TNFSF18 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFSF18 | |
Human recombinant GITR Ligand/TNFSF18 (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0312-4Q25 | |
Flow Cytometry | |
Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Mouse | |
Protein G purified | |
Apoptosis | |
8995 | |
Store at 4°C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title