Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex Safety Products CorXcel Micro-Foam Nitrile Gloves with HPPE Liner - GL603C5 Series
Pyramex Safety CorXcel Micro-Foam Nitrile Gloves are tear and abrasion resistant
Manufacturer: Pyramex Safety Products GL603C5XL
Description
- 13 gauge cut-resistant HPPE liner
- Optional black color does not show dirt or grime
- Foam nitrile absorbs oil to increase grip
- Additional foam coating acts like a sponge, helps increase gripping power
- ANSI Cut Level: 4
- Abrasion Rating: 4
- Circular Blade Cut Rating: 5
- Tear Rating: 4
- Puncture Rating: 3
Specifications
Foam Nitrile | |
ASTM F2878 LEVEL 3 | |
Black |
HPPE | |
X-Large | |
ANSI 4 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title