Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glacial Acetic Acid, USP, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O

AC110, 64-19-7, C2H4O2

Spectrum™ Glacial Acetic Acid, USP does have some antibacterial and antifungal properties but is used primarily as a pH adjuster. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN2789
Percent Purity 99.5 to 100.5%
Packaging Amber Glass Bottle
Quantity 500 mL