Learn More
Selleck Chemical LLCSupplier Diversity Partner Glasdegib (PF-04449913) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S71605MG
Glasdegib (PF-04449913) is a potent, and orally bioavailable Smoothened (Smo) inhibitor with IC50 of 5 nM. Phase 2.
Catalog No. 50-194-8095