Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foss™ Glass Bottles for Sample Mills

Accessory for use with CM 290 Cemotec, CT 293 Cyclotec, KN 295 Knifetec Sample Mills.

$35.25 - $239.00

Specifications

Product Type Bottle
Material Glass
Sample Volume (Metric) 350mL
For Use With (Equipment) CM 290 Cemotec, CT 293 Cyclotec, KN 295 Knifetec Sample Mills
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
12-053-132
View Documents
Foss™
60068548
4/Pk.
Pack of 4 for $35.25
Add to cart
 
12-053-133
View Documents
Foss™
60076311
40/Pk.
Pack of 40 for $239.00
Add to cart
 
Description

Description

  • For use with CM 290 Cemotec, CT 293 Cyclotec, KN 295 Knifetec Sample Mills
  • Volume: 350mL
Specifications

Specifications

Bottle
Glass
350mL
CM 290 Cemotec, CT 293 Cyclotec, KN 295 Knifetec Sample Mills
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit