Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Glass Buchner Funnel - Plain Stem
Glass Buchner funnels with plain stems and G-3 sintered disks
Supplier: Eisco™ CH0466D
Description
- Made from high quality borosilicate glass
- Includes G-3 sintered disk (15-40 microns)
- Plain Stem
- Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
9 cm | |
Borosilicate Glass | |
Yes | |
filtering liquids | |
G-3 |
500 mL | |
17.8 cm | |
Clear | |
1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction