Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Glass-Fiber Frit

For use with 15 mm I.D. Tubes

Manufacturer:  MilliporeSigma™ Supelco™ 21537U

Catalog No. 11-101-0702


Add to cart

Description

Description

Specifications

Specifications

15 mm I.D. Tubes
Glass-Fiber
50/Pk.
Frit
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit