Learn More
Scientific Cell Company Glass Filter for Testing Wavelength and Photometric Accuracy
This glass filter is used for testing the wavelength accuracy and photometric accuracy (0.5 to 1 Abs).
Manufacturer:  Scientific Cell Company 666F7
Catalog No. 14-397-509
Add to cart
Description
Description
  • Wavelength accuracy certified at: wavelengths: 329, 472, 512, 681, 875nm; slit width: 1nm
  • Photometric accuracy certified at: wavelengths: 270, 280, 297, 320, 340nm;  slit width: 1nm; serial no.
  • In high-quality storage case with DAkkS calibration certificate
  • Light path:  Didymium            
Specifications
Specifications
Glass, Solid
Individual filter