Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Cell Company Glass Filters for Testing Photometric Accuracy

For testing the photometric accuracy.

Manufacturer:  Scientific Cell Company 666F4

 View more versions of this product

Catalog No. 14-397-507


Add to cart

Includes: DAkkS calibration certificate

Description

Description

  • Certified at wavelengths: 440, 465, 546.1, 590, 635nm slit width
  • 1nm serial no.
  • Type: Solid
  • Individual filter
  • Light Path: Neutral Density
  • Packed in high-quality storage case
Specifications

Specifications

Testing photometric accuracy (1.0 Abs)
Glass Filter
DAkkS calibration certificate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit