Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Glass Flushing Adapter

Adapters include a 2 mm, T-bore PTFE stopcock and enable vessels to be purged with an inert gas.

Manufacturer:  Synthware™ A681922G

 View more versions of this product

Catalog No. 31-502-401


Add to cart

Description

Description

  • Flushing adapters enable vessels to be purged with an inert gas
  • Includes a 2 mm, T-bore PTFE stopcock
Specifications

Specifications

Adapter
Borosilicate Glass 3.3
93°C
Class A
Storage/Reaction Vessels or Flasks
19/22
2 mm
Autoclavable
200°F
Flushing/Filling Attached Vessels with an Inert Gas
Glass T-Bore Stopcock
Hose
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit