Learn More
DWK Life Sciences Wheaton™ Glass Jug, Clear, 2000mL
Glass jugs made of clear Type III borosilicate/soda-lime glass with attached screw caps.
Manufacturer:  DWK Life Sciences W220991
 View more versions of this product
Catalog No. 03-084-169
  Check Availability
Add to cart
Description
Description
Caps are available in a variety of materials to meet your compatibility needs.
Specifications
Specifications
2000mL
PE Cone
Type III Borosilicate Glass
Glass Jug
Phenolic
Clear
6/Case
Black