Learn More
Whatrman™ Glass Microfiber Filter Paper, 93AH Pre-Rinsed and Dried: PROMO
Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Sheets are also used in cell harvesting, liquid scintillation, and monitoring pollution.
Manufacturer:  Whatrman™ 2827047PM
Catalog No. 45999003LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Smooth surface for efficient precipitate recovery
  • Nominal particle retention of 1.5µm
  • Withstands temperatures up to 500°C
  • Fast filtration speed
Specifications
Specifications
3.7 sec./100mL/in.2
934-AH
1.5μm