Learn More
DWK Life Sciences Wheaton™ Glass Pasteur Pipettes
Disposable glass Pasteur Pipettes made from Type 1 clear glass.
Manufacturer:  DWK Life Sciences W357331
 View more versions of this product
Catalog No. 02-913-363
  Check Availability
Add to cart
Description
Description
  • Used for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method described in the 21st Edition (2005) of Standard Methods for the Examination of Water and Wastewater, Method 3500-As B
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
  • Assembled apparatus is freestanding and easy to use
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Specifications
Clear
Arsenic analysis
Assembled
Borosilicate Glass
Yes
250mm
Pasteur Pipettes