Learn More
Leica Microsystems 3P GLASS PEN MEMBRANE SLD, 50/CS
Manufacturer:  Leica Microsystems 3P 11505158
Encompass
Glass slides with PEN foil membrane, 26 mm x 76 mm, 50 per box. PEN (poly ethylene napthalate) membrane is 2.0 microns thick. Used for Laser Microdissection.
Catalog No. NC0496333