Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™: PROMO
Vial File provides a compact, easily accessible means of sample storage
$82.79 - $159.45
Specifications
Autoclavable Yes
Type Glass Sample Vial
For Use With (Application) Used for storing and transporting vials
Thread Style Screw
View More Specs
Products 8
Catalog Number Mfr. No. Volume (Metric) Volume (Metric) Graduated Liner Material Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Volume (Metric) Graduated Liner Material Price Quantity & Availability  
11-302-8PM View Documents DWK Life Sciences
224952PM
4.0 mL No PTFE/Silicone Case of 40 for $145.51
Add to cart
02-912-502PM View Documents DWK Life Sciences
W224896PM
2.0 mL No 14B Rubber Case of 60 for $123.72
Add to cart
02-912-509PM View Documents DWK Life Sciences
W224693PM
2.0 mL No PTFE/14B Rubber Case of 60 for $126.33
Add to cart
02-912-510PM View Documents DWK Life Sciences
W224694PM
4.0 mL No PTFE/14B Rubber Case of 40 for $99.31
Add to cart
02-912-511PM View Documents DWK Life Sciences
W224695PM
2.0 mL No PTFE/14B Rubber Case of 40 for $135.05
Add to cart
03-336-PM View Documents DWK Life Sciences
224891PM
2.0 mL No 14B Rubber Case of 60 for $100.19
Add to cart
03-336-5PM View Documents DWK Life Sciences
224892PM
4.0 mL No 14B Rubber Case of 40 for $82.79
Add to cart
03-336-6PM View Documents DWK Life Sciences
224950PM
1.5 mL No PTFE/Silicone Case of 60 for $159.45
Add to cart
Description
Description
Caps attached to vials.
  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.
Specifications
Specifications
Yes
Glass Sample Vial
Used for storing and transporting vials
Screw