Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Eisco™ Glass Test Tubes with Screw Caps Products for Science Education
Glass test tubes with screw caps are useful for a variety of laboratory applications and ensure a leak proof seal.
Specifications
Autoclavable Autoclavable
Closure Screw Cap
Closure Material Bakelite
Color Clear
Graduated No
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S15064 Products for Science Education
View Documents
Eisco™
CH0724APK24
5 mL
Each for $30.50
N/A
S15065 Products for Science Education
View Documents
Eisco™
CH0724CPK24
20 mL
Each for $43.75
N/A
S15066 Products for Science Education
View Documents
Eisco™
CH0724DPK24
25 mL
Each for $45.75
N/A
Description
Description
  • Made of high quality, heavy duty Borosilicate 3.3 glass
  • Fitted with a Bakelite rubber-lined screw cap to ensure a leak proof seal
  • Low temperature gradient, capable of withstanding high temperatures & thermal shocks
  • Superior chemical durability
  • Plain, round bottom design
  • Autoclavable
Specifications
Specifications
Autoclavable
Bakelite
No
Round Bottom
Screw Cap
Clear
Borosilicate Glass 3.3
Non-sterile