Alfa Aesar™ Glassclad™ 18

Manufacturer: Alfa Aesar™ 43293-AC

Quantity 25mL

 View more versions of this product

Catalog No. AA43293AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00240276
    UN Number UN1993
    Chemical Name or Material Glassclad™ 18

    Specifications

    Density 0.90g/mL
    MDL Number MFCD00240276
    UN Number UN1993
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H332-H319
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off imm
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 10°C (50°F)
    Physical Form Liquid
    Quantity 25mL
    Chemical Name or Material Glassclad™ 18

    Description

    • As organosilane concentrate which, when properly applied to glass, provides a surface which is water repellent, has improved lubricity, and surface resistivity

    GHS H Statement
    H225-H332-H319
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off imm

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit