Learn More
Alfa Aesar™ Glassclad™ 18
Manufacturer:  Alfa Aesar™ 43293-AE
Quantity 100mL
 View more versions of this product
Catalog No. AA43293AE
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • As organosilane concentrate which, when properly applied to glass, provides a surface which is water repellent, has improved lubricity, and surface resistivity
Description & Specifications
Chemical Identifiers
MFCD00240276
UN1993
Glassclad™ 18
Specifications
100mL
0.90g/mL
UN1993
Glassclad™ 18
Liquid
MFCD00240276
10°C (50°F)
Safety and Handling
GHS H Statement
H225-H332-H319
GHS P Statement
P210-P261-P280-P240-P241-P303+P361+P353-P305+P351+P338-P304+P340-P312-P501a
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.