Learn More
Electron Microscopy SciencesSupplier Diversity Partner Enlarger Glassless Carrier, 6 x 4.5 cm
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 7487322
Encompass_Preferred
All aluminum precision carriers with hinged sandwich design for fast handling and loading. Guide pins position negatives in line with carrier aperture. Offset tabs permits rapid opening. Can be used on Omega D5 Enlargers.
Size: 6 x 4.5 cm
Catalog No. 50-250-79