Learn More
Alfa Aesar™ Glassy Carbon Conical Crucible
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Manufacturer:  Alfa Aesar™ 39003KT
 View more versions of this product
Catalog No. AA39003KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Glassy Carbon Crucible
Top: 31mm; Bottom: 17mm
Conical
10mL
27mm
Glassy Carbon