Learn More
Alfa Aesar™ Glassy Carbon Cylindrical Crucible
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Manufacturer:  Alfa Aesar™ 39011KT
 View more versions of this product
Catalog No. AA39011KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Glassy Carbon Crucible
40mm
Cylindrical
50mL
56mm
Glassy Carbon