Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glassy Carbon Lid;Top Dia (mm), 92;Bottom Dia (mm), 72, Alfa Aesar™
Click to view available options
Quantity:
1 ea.
Packaging:
Shrink-wrapped
Specifications
Packaging | Shrink-wrapped |
Quantity | 1 ea. |
Chemical Name or Material | Glassy Carbon Lid;Top Dia (mm) |
Safety and Handling
TSCA : U
Recommended Storage : Ambient temperatures