Learn More
Glassy carbon plate, 1mm (0.04in) thick, type 1, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
Supplier: Thermo Scientific Chemicals 038024FI
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
Tube | |
MFCD00133992 |
7440-44-0 | |
50 x 50 mm | |
Insoluble. |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only