Learn More
Glassy carbon plate, 1mm (0.04in) thick, type 2, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloy, powder and glass. Glassy carbon plate is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices. It also used in solar energy applications.
$400.00 - $1520.65
Specifications
Chemical Name or Material | Glassy carbon plate |
---|---|
CAS | 7440-44-0 |
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA38021FI
|
Thermo Scientific Chemicals
038021FI |
50 x 50 mm |
Each for $400.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38021GH
|
Thermo Scientific Chemicals
038021GH |
100 x 100 mm |
Each
for $1,520.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Used in casting, smelting of metal, hard alloy, powder and glass. Glassy carbon plate is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices. It also used in solar energy applications. It offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
Tube | |
Insoluble. |
7440-44-0 | |
MFCD00133992 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.