Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon plate, 2mm (0.08in) thick, type 1, Thermo Scientific Chemicals

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.

Specifications

Chemical Name or Material Glassy carbon plate
Packaging Tube
MDL Number MFCD00133992
Solubility Information Insoluble in water.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA38025FI
View Documents
Thermo Scientific Chemicals
038025FI
50 x 50 mm
Each for $330.00
Order in Bulk

Request bulk or custom formats

N/A
AA38025GH
View Documents
Thermo Scientific Chemicals
038025GH
100 x 100 mm
Each for $1,057.00
Order in Bulk

Request bulk or custom formats

N/A
Description

Description

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glassy carbon plate
Tube
MFCD00133992
Insoluble in water.
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit