Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glassy carbon plate, 4mm (0.16 in.) thick, type 1, Alfa Aesar
Manufacturer: Alfa Aesar 38026FF
Specifications
Glassy carbon plate | |
25 x 25mm | |
MFCD00133992 | |
≈3.8g/25x25mm |
Tube | |
7440-44-0 | |
Insoluble in water. |
Safety and Handling
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title