Learn More
Glassy carbon plate, 4mm (0.16in) thick, type 1, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038026FI
Description
Glassy carbon plate, is used as fuel cell and Due to their specific surface orientation, glassy carbon is employed as an electrode material for the fabrication of sensors. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments. Chemically modified glasssy electrodes have been employed for the analysis of organic molecules (viz., Paracetamol, aspirin, caffeine, phenol, catechol, resorcinol, hydroquinone, dopamine, L-dopa, epinephrine, nor epinephrine, methyl parathion, ethyl parathion, venlafaxine, desvenlafaxine, imipramine, trimipramine, desipramine etc.) as well as metal ions (bismuth, antimony etc.)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
Tube | |
MFCD00133992 |
7440-44-0 | |
50 x 50 mm | |
Insoluble in water. |
Safety and Handling
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only