Learn More
Glassy carbon plate, 4mm (0.16in) thick, type 2, Thermo Scientific Chemicals
Used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.
$178.65 - $1160.65
Specifications
Chemical Name or Material | Glassy carbon plate |
---|---|
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA38023FF
|
Thermo Scientific Chemicals
038023FF |
25 x 25 mm |
Each
for $178.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38023FI
|
Thermo Scientific Chemicals
038023FI |
50 x 50 mm |
Each
for $375.65
Save 8% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38023GH
|
Thermo Scientific Chemicals
038023GH |
100 x 100 mm |
Each
for $1,160.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Glassy carbon plate, is used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
Tube | |
MFCD00133992 | |
Insoluble in water. |
Safety and Handling
EINECSNumber : 231-153-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.