Learn More
Glassy carbon plate, 4mm (0.16in) thick, type 2, Thermo Scientific Chemicals
Used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.
Supplier: Thermo Scientific Chemicals 038023GH
Description
Glassy carbon plate, is used as fuel cell and other electrochemical electrodes. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
100 x 100 mm | |
Insoluble in water. |
Tube | |
MFCD00133992 |
Safety and Handling
EINECSNumber : 231-153-3
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only