Learn More
Glassy carbon rod, 1mm (0.04in) dia, type 2, Thermo Scientific Chemicals
Widely used as a electrode material in electrochemistry.
Supplier: Thermo Scientific Chemicals 037996DR
Description
Due to its distinctive property, it is widely used as a electrode material in electrochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
MFCD00133992 |
7440-44-0 | |
200 mm | |
Insoluble. |
Safety and Handling
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only