Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon rod, 2mm (0.08 in.) dia, type 2, Alfa Aesar™

Widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.

$136.10 - $421.50

Specifications

MDL Number MFCD00133992
Chemical Name or Material Glassy carbon rod
Packaging Tube
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA37997DM View Documents Alfa Aesar
37997DM
100mm Each for $220.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA37997DR View Documents Alfa Aesar
37997DR
200mm Each for $421.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA37997DJ View Documents Alfa Aesar
37997DJ
50mm Each for $136.10
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

MFCD00133992
Glassy carbon rod
Tube
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit