Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon rod, 2mm (0.08in) dia, type 1, Thermo Scientific™

Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

$144.66 - $484.23

Specifications

MDL Number MFCD00133992
Chemical Name or Material Glassy carbon rod
Packaging Tube
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA38010DJ
View Documents
Thermo Scientific Chemicals
38010DJ
50mm
Each for $144.66
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA38010DR
View Documents
Thermo Scientific Chemicals
38010DR
200mm
Each for $484.23
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA38010DM
View Documents
Thermo Scientific Chemicals
38010DM
100mm
Each for $255.95
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

MFCD00133992
Glassy carbon rod
Tube
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit